EMAIL THIS PAGE TO A FRIEND
Developmental dynamics : an official publication of the American Association of Anatomists
Halogenated nucleotide labeling of nascent RNAs reveals dynamic transcription in growing pig oocytes.
PMID 23161795
Abstract
Germ cells differentiate into oocytes in females and are arrested at the first meiotic prophase. However, during arrest, oocytes undergo a growth phase leading to a dramatic increase in size, which is under control of transcription events. In the current study, we examined the transcriptional activity of growing pig oocytes using an immunocytochemical approach. Our data showed that fluorouridine (FU), a halogenated nucleotide, can be successfully incorporated into synthesizing RNAs and detected using a specific monoclonal antibody. Using this method, we identified dynamic changes in transcriptional activity patterns in growing pig oocytes. Oocytes obtained from small follicles exhibited the highest level of transcription, while at the final phase of growth, transcription was no longer detected. These transcriptional changes were concomitant with chromatin compaction resulting in a tightly packed ring-like chromatin conformation surrounding the nucleolar structure. Also, FU incorporation appeared sensitive to the biochemical manipulation of transcription, because transcriptional inhibitors induced a decrease in signal intensity from FU labeling and transcriptional activation caused an increase in FU signal intensity. Our data collectively support that a direct link exists between chromatin configuration and transcriptional activity in pig oocytes, and support the suitability of FU for studies on transcription-related events in mammalian oocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5130
5-Fluorouridine, proapoptotic anitproliferative plant growth regulator
C9H11FN2O6