EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Treatment of contagious bovine pleuropneumonia.
PMID 23161933
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33700
Danofloxacin, VETRANAL, analytical standard
C19H20FN3O3