EMAIL THIS PAGE TO A FRIEND
Clinical laboratory
Maternal plasma advanced glycation end products concentrations in response to oral 50-gram glucose load in mid-pregnancy: a pilot study.
PMID 23163122
Abstract
Accumulating evidence documents the initiation of diverse physiologic and biochemical responses subsequent to an oral glucose load. We sought to evaluate the extent to which acute hyperglycemia, resulting from a 50-gram glucose load, contributes to changes in maternal plasma concentrations of advanced glycation end products (AGEs), a heterogeneous group of molecules formed from the non-enzymatic reaction of reducing sugars with free amino groups of proteins, lipids, and nucleic acids. Blood specimens were collected from each participant in mid-pregnancy using standard procedures before and after a 50-gram oral glucose load. Maternal plasma methylglyoxal (MG), pentosidine and N(epsilon)-(carboxymethyl)lysine (CML) (free and bound) were measured by HPLC-MS/MS method. Non-parametric methods were employed for statistical analysis. Median plasma MG increased 1.27 fold as a result of acute hyperglycemia. Median bound CML concentrations were elevated 21% in post-load plasma samples as compared with pre-load samples, while median free pentosidine concentrations were 51% lower (both p-values < 0.05). Future studies of larger populations and longer periods of follow-up are warranted to investigate the consequences of acute and chronic hyperglycemia on placental function and fetal development.