EMAIL THIS PAGE TO A FRIEND
CNS neuroscience & therapeutics
Effects of oxymatrine on the neuropathic pain induced by chronic constriction injury in mice.
PMID 23164255
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N142
NO-711 hydrochloride, ≥98% (HPLC)
C21H22N2O3 · HCl