EMAIL THIS PAGE TO A FRIEND
Annales francaises d'anesthesie et de reanimation
[Phaeochromocytoma revealed by a cardiogenic shock treated by extracorporeal life support].
PMID 23164653
Abstract
Phaeochromocytoma is a catecholamine-secreting tumour that originates from the chromaffin cells in the adrenal medulla in 85% of the cases. Phaeochromocytoma typically presents with the classic signs and symptoms of paroxysmal hypertension, tachycardia, and episodic headache in young adults. However, it rarely may manifest as cardiogenic shock due to a catecholamine induced cardiomyopathy. We report the use of central extracorporeal life support (ECLS) in a young man admitted to our department because of cardiogenic shock caused by phaeochromocytoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U100
Urapidil hydrochloride, solid
C20H29N5O3 · HCl