EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Detection of small differences in the hydrophilicity of ions using the LCST-type phase transition of an ionic liquid-water mixture.
PMID 23164982
Abstract
The phase separation temperature of the tetrabutylphosphonium trifluoroacetate-water mixture, which undergoes a lower critical solution temperature-type phase transition, is highly sensitive to the hydrophilicity of added salts; this system can be used to compare the hydrophilicity of the added ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
189138
Tetrabutylphosphonium bromide, 98%
C16H36BrP
144800
Tetrabutylphosphonium chloride, 96%
C16H36ClP