EMAIL THIS PAGE TO A FRIEND
Interactive cardiovascular and thoracic surgery
eComment. The problem of anaemia correction in cardiac surgery patients.
PMID 23166216
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5381
Iron(II) fumarate, 97%
C4H2FeO4