EMAIL THIS PAGE TO A FRIEND
Dento maxillo facial radiology
Synovial chondromatosis of the temporomandibular joint with calcium pyrophosphate dihydrate crystal deposition disease (pseudogout).
PMID 23166363
Abstract
This report describes a very rare case of synovial chondromatosis with deposition of calcium pyrophosphate dihydrate (CPPD) crystals (pseudogout) in the temporomandibular joint (TMJ) of a 46-year-old male patient. Synovial chondromatosis is a non-neoplastic disease characterized by metaplasia of the connective tissue leading to chondrogenesis in the synovial membrane. Pseudogout is an inflammatory disease of the joints caused by the deposition of CPPD, producing similar symptoms to those observed in gout but not hyperuricaemia. Both diseases commonly affect the knee, hip and elbow joints, but rarely affect the TMJ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
693871
Calcium phosphate, amorphous, nanopowder, <150 nm particle size (BET)