PloS one
Biomarkers of whale shark health: a metabolomic approach.
PMID 23166652
Abstract
In a search for biomarkers of health in whale sharks and as exploration of metabolomics as a modern tool for understanding animal physiology, the metabolite composition of serum in six whale sharks (Rhincodon typus) from an aquarium collection was explored using (1)H nuclear magnetic resonance (NMR) spectroscopy and direct analysis in real time (DART) mass spectrometry (MS). Principal components analysis (PCA) of spectral data showed that individual animals could be resolved based on the metabolite composition of their serum and that two unhealthy individuals could be discriminated from the remaining healthy animals. The major difference between healthy and unhealthy individuals was the concentration of homarine, here reported for the first time in an elasmobranch, which was present at substantially lower concentrations in unhealthy whale sharks, suggesting that this metabolite may be a useful biomarker of health status in this species. The function(s) of homarine in sharks remain uncertain but it likely plays a significant role as an osmolyte. The presence of trimethylamine oxide (TMAO), another well-known protective osmolyte of elasmobranchs, at 0.1-0.3 mol L(-1) was also confirmed using both NMR and MS. Twenty-three additional potential biomarkers were identified based on significant differences in the frequency of their occurrence between samples from healthy and unhealthy animals, as detected by DART MS. Overall, NMR and MS provided complementary data that showed that metabolomics is a useful approach for biomarker prospecting in poorly studied species like elasmobranchs.