EMAIL THIS PAGE TO A FRIEND
Hand clinics
Trapezium resection with mersilene suspension sling.
PMID 23168026
Abstract
The reason there are numerous techniques for thumb carpometacarpal arthroplasty is that none of them are perfect. Sometimes the simplest procedures work best. This article presents a simple alternative, using a readily available suture to stabilize the thumb after resection of the trapezium, with long-term success.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
429252
Poly(ethylene terephthalate), granular