EMAIL THIS PAGE TO A FRIEND
Hand clinics
A carpal ligament substitute part 1: polyester suture.
PMID 23168035
Abstract
We have searched for a synthetic substitute for the carpal ligaments, which would be widely available and easy to use. Four loops of 2-0 polyester fiber suture (Mersilene) were found to exceed the ultimate tensile strength of the scapholunate interosseous ligament. This construct approximates a normal ligament stress/strain curve and can theoretically facilitate fibrous tissue ingrowth. It is readily available, easy to handle, and inexpensive. Based on these findings, we recommend the use of polyester suture in the reconstruction of carpal and other ligaments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
429252
Poly(ethylene terephthalate), granular