EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of phenanthridinones via palladium-catalyzed C(sp2)-H aminocarbonylation of unprotected o-arylanilines.
PMID 23168677
Abstract
An efficient synthesis of free (NH)-phenanthridinones through Pd-catalyzed C(sp(2))-H aminocarbonylation of unprotected o-arylanilines under an atmospheric pressure of CO has been developed. Some ortho heteroarene substituted anilines as well as N-alkyl protected o-arylanilines are also suitable substrates for this C-H aminocarbonylation reaction.