EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
A linear allergic contact dermatitis to p-tert-butylphenol formaldehyde resin sectorially present in a neoprene orthopedic brace: role of spectroscopy.
PMID 23169214