EMAIL THIS PAGE TO A FRIEND
Nihon Eiseigaku Zasshi. Japanese Journal of Hygiene
[Effects of environmental temperatures on the cadmium-induced testicular injury (author's transl)].
PMID 231694
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-537
Anti-FAK Antibody, clone 4.47, clone 4.47, Upstate®, from mouse