EMAIL THIS PAGE TO A FRIEND
Journal of clinical biochemistry and nutrition
Identification of the radicals formed in the reactions of some endogenous photosensitizers with oleic acid under the UVA irradiation.
PMID 23170043
Abstract
Electron spin resonance measurements were performed for the reactions of some endogenous photosensitizers (flavin mononucleotide or flavin adenine dinucleotide or folic acid or β-nicotinamide adenine dinucleotide or β-nicotinamide adenine dinucleotide phosphate or pyridoxal-5'-phosphate or urocanic acid) with oleic acid under the ultraviolet light A irradiation using α-(4-pyridyl-1-oxide)-N-tert-butylnitrone as a spin trap reagent. Of the endogenous photosensitizers, prominent electron spin resonance signals (α(N)xa0=xa01.58xa0mT and α(H)βxa0=xa00.26xa0mT) were observed for the reaction mixture of flavin mononucleotide (or flavin adenine dinucleotide or folic acid), suggesting that radical species form in the reaction mixtures. Singlet oxygen seems to participate in the formation of the radicals because the electron spin resonance peak heights increased for the reactions in D(2)O to a great extent. A high performance liquid chromatography-electron spin resonance-mass spectrometry was employed to identify the radicals formed in the reactions of the endogenous photosensitizers (flavin mononucleotide or flavin adenine dinucleotide or folic acid) with oleic acid under the ultraviolet light A irradiation. The high performance liquid chromatography-electron spin resonance-mass spectrometry analyses showed that 7-carboxyheptyl and 1-(3-carboxypropyl)-4-hydroxybutyl radicals form in the reaction mixture of flavin mononucleotide (or flavin adenine dinucleotide or folic acid).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459119
2,2,6,6-Tetramethyl-4-piperidone, 95%
C9H17NO
859796
4-Imidazoleacrylic acid, 99%
C6H6N2O2