EMAIL THIS PAGE TO A FRIEND
Organic letters
Lewis acid catalyzed intramolecular condensation of ynol ether-acetals. Synthesis of alkoxycycloalkene carboxylates.
PMID 23170869
Abstract
Treatment of ynol ether-tethered dialkyl acetals with catalytic quantities of scandium triflate in CH(3)CN gives rise to five-, six-, and seven-membered alkoxycycloalkene carboxylates in good to excellent yields. Tri- and tetrasubstituted carbocyclic and heterocyclic alkenes may be formed by this method, and the products obtained may serve as useful intermediates for natural product synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261262
Scandium, powder, 99.9% trace rare earth metals basis
Sc