EMAIL THIS PAGE TO A FRIEND
BMC urology
Resistance profiles of urinary tract infections in general practice--an observational study.
PMID 23171154
Abstract
Guideline recommendations on therapy in urinary tract infections are based on antibiotic resistance rates. Due to a lack of surveillance data, little is known about resistance rates in uncomplicated urinary tract infection (UTI) in general practice in Germany. In a prospective observational study, urine cultures of all women presenting with urinary tract infections in general practice were analysed. Resistance rates against antibiotics recommended in German guidelines on UTI are presented. In a prospective, multi-center observational study general practitioner included all female patients ≥ 18 years with clinically suspected urinary tract infection. Only patients receiving an antibiotic therapy within the last two weeks were excluded. 40 practices recruited 191 female patients (mean age 52 years; range 18-96) with urinary tract infections. Main causative agent was Escherichia coli (79%) followed by Enterococcus faecalis (14%) and Klebsiella pneumoniae (7.3%).Susceptibility of E.coli as the main causative agent was highest against fosfomycin and nitrofurantoin, with low resistance rates of 4,5%; 2,2%. In 17,5%, E.coli was resistant to trimethoprim and in 8,5% to ciprofloxacin. Resistance rates of uropathogens from unselected patients in general practice differ from routinely collected laboratory data. These results can have an impact on antibiotic prescribing and treatment recommendations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1191
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H6N4O5