EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Benzyl isothiocyanate induces protective autophagy in human prostate cancer cells via inhibition of mTOR signaling.
PMID 23172666
Abstract
Benzyl isothiocyanate (BITC) is a dietary chemopreventive agent that inhibits the growth of various human cancer cells by causing apoptotic cell death. In this study, we demonstrate that BITC not only induces apoptosis but also induces autophagy in human hormone-sensitive (Rv1) and -refractory (PC3) prostate cancer cells. In BITC-treated cells, the induction of autophagy was detected by monitoring the processing of an autophagy marker protein, microtubule-associated protein 1 light chain 3 (LC3), the aggregation of LC3 into granular structures and the formation of acidic organelles. Inhibition of autophagy using 3-methyladenine increased BITC-induced apoptosis, whereas the administration of caspase inhibitor suppressed BITC-induced cell death. Our data also showed that BITC inhibits mammalian target of rapamycin (mTOR) kinase activity in a dose-dependent manner. The expression of phospho-mTOR (Ser2481), an indicator of mTOR intrinsic catalytic activity, and phospho-UNC-51-like kinase 1 (Ser757), a direct substrate of mTOR, were decreased in BITC-treated cells. However, the increased expression of phospho-mTOR (Ser2448), phospho-AKT (Ser473) and antiapoptotic Bcl-2 were detected only in PC3 cells at later stages of BITC treatment. Collectively, our results show that BITC induces a protective autophagy response in Rv1 and PC3 cells through inhibition of the mTOR signaling pathway. Activation of the AKT survival pathway was only observed in PC3 cells, representing a resistance mechanism of advanced prostate cancer upon BITC treatment. These findings could potentially contribute to the beneficial effect of BITC in prostate cancer treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252492
Benzyl isothiocyanate, 98%
C8H7NS
W442800
Benzyl isothiocyanate, 98%, FG
C8H7NS