EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Comparison of splicing factor 3b inhibitors in human cells.
PMID 23172726
Abstract
Name your splice: FR901464 analogues and herboxidiene inhibit constitutive splicing, most likely by inhibiting spliceosomal subunit SF3b. A parallel comparison of these compounds in a cell-based assay system showed meayamycin B as the most potent splicing inhibitor among these small molecules.