EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Isoprenyl-thiourea and urea derivatives as new farnesyl diphosphate analogues: synthesis and in vitro antimicrobial and cytotoxic activities.
PMID 23174318
Abstract
A series of new isoprenyl-thiourea and urea derivatives were synthesized by the reaction of alkyl or aryl isothiocyanate or isocyanate and primary amines. The structures of the compounds were established by (1)H NMR, (13)C NMR, MS, HRMS and elemental analysis. The new compounds were screened for in vitro antimicrobial activity against seven strains representing different types of gram-positive and gram-negative bacteria. More than a third of the synthesized compounds showed variable inhibition activities against the tested strains. Best antimicrobial activities were found for those thiourea analogues with 3-methyl-2-butenyl, isobutyl or isopentyl groups and aromatic rings possessing electron withdrawing substituents. The new compounds were also subjected to a preliminary screening for antitumoral activity. The presence of a highly lipophilic group and an electron withdrawing group in the aromatic rings enhanced anticancer activity of the synthesized compounds, showing in most cases more activity than that of the controls.