EMAIL THIS PAGE TO A FRIEND
Science translational medicine
Modulation of vigilance in the primary hypersomnias by endogenous enhancement of GABAA receptors.
PMID 23175709
Abstract
The biology underlying excessive daytime sleepiness (hypersomnolence) is incompletely understood. After excluding known causes of sleepiness in 32 hypersomnolent patients, we showed that, in the presence of 10 μM γ-aminobutyric acid (GABA), cerebrospinal fluid (CSF) from these subjects stimulated GABA(A) receptor function in vitro by 84.0 ± 40.7% (SD) relative to the 35.8 ± 7.5% (SD) stimulation obtained with CSF from control subjects (Student's t test, t = 6.47, P < 0.0001); CSF alone had no effect on GABA(A) signaling. The bioactive CSF component had a mass of 500 to 3000 daltons and was neutralized by trypsin. Enhancement was greater for α2 subunit- versus α1 subunit-containing GABA(A) receptors and negligible for α4 subunit-containing ones. CSF samples from hypersomnolent patients also modestly enhanced benzodiazepine (BZD)-insensitive GABA(A) receptors and did not competitively displace BZDs from human brain tissue. Flumazenil--a drug that is generally believed to antagonize the sedative-hypnotic actions of BZDs only at the classical BZD-binding domain in GABA(A) receptors and to lack intrinsic activity--nevertheless reversed enhancement of GABA(A) signaling by hypersomnolent CSF in vitro. Furthermore, flumazenil normalized vigilance in seven hypersomnolent patients. We conclude that a naturally occurring substance in CSF augments inhibitory GABA signaling, thus revealing a new pathophysiology associated with excessive daytime sleepiness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6300
Flumazenil, >99% (HPLC), solid
C15H14FN3O3