EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
The benzylidenecarbene-phenylacetylene rearrangement: an experimental and computational study.
PMID 23176163
Abstract
Benzylidenecarbene was generated from a new photochemical source, 1-benzylidene-1a,9b-dihydro-1H-cyclopropa[l]phenanthrene, in deuterated benzene at ambient temperature. The carbene undergoes a facile rearrangement to phenylacetylene and could not be trapped by olefins. Generation of the carbene bearing a (13)C label at the β-carbon produced phenylacetylene in which the label was found exclusively at the carbon adjacent to the phenyl ring. This overwhelming preference for H shift is consistent with B3LYP and CCSD(T) calculations. The label distribution observed in this work, however, contrasts previously reported high-temperature flash vacuum pyrolysis results where the interconversion of carbene and alkyne leads to the scrambling of labels over both alkynyl (sp) carbons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
117706
Phenylacetylene, 98%
C8H6