EMAIL THIS PAGE TO A FRIEND
International journal of experimental pathology
Changes of imidazoline receptors in spontaneously hypertensive rats.
PMID 23176371
Abstract
The role of imidazoline receptors in the regulation of vascular function remains unclear. In this study, we evaluated the effect of agmatine, an imidazoline receptor agonist, on systolic blood pressure (SBP) in spontaneously hypertensive rats (SHRs) and investigated the expressions of imidazoline receptors by Western blot. The isometric tension of aortic rings isolated from male SHRs was also estimated. Agmatine decreased SBP in a dose-dependent manner in SHRs but not in the normal group [Wistar-Kyoto (WKY) rats]. This reduction in SBP in SHRs was abolished by BU224, a selective antagonist of imidazoline I(2) -receptors. Higher expression of imidazoline receptors in SHR was observed. Moreover, agmatine-induced relaxation in isolated aortic rings precontracted with phenylephrine or KCl. This relaxation was also abolished by BU224 but was not modified by efaroxan, an imidazoline I(1) -receptor antagonist. Agmatine-induced relaxation was also attenuated by PNU 37883, a selective blocker of vascular ATP-sensitive potassium (K(ATP) ) channels. Additionally, vasodilatation by agmatine was reduced by an inhibitor of protein kinase A (PKA). We suggest that agmatine can lower blood pressure in SHRs through activation of the peripheral imidazoline I(2) -receptor, which is expressed more highly in SHRs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3263
Efaroxan hydrochloride, ≥98% (HPLC)
C13H16N2O · HCl