EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Electrochemical communication between microbial cells and electrodes via osmium redox systems.
PMID 23176477
Abstract
Electrochemical communication between micro-organisms and electrodes is the integral and fundamental part of BESs (bioelectrochemical systems). The immobilization of bacterial cells on the electrode and ensuring efficient electron transfer to the electrode via a mediator are decisive features of mediated electrochemical biosensors. Notably, mediator-based systems are essential to extract electrons from the non-exoelectrogens, a major group of microbes in Nature. The advantage of using polymeric mediators over diffusible mediators led to the design of osmium redox polymers. Their successful use in enzyme-based biosensors and BFCs (biofuel cells) paved the way for exploring their use in microbial BESs. The present mini-review focuses on osmium-bound redox systems used to date in microbial BESs and their role in shuttling electrons from viable microbial cells to electrodes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263257
Osmium, powder, 99.9% trace metals basis
Os