EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Metabolic studies of the Amaryllidaceous alkaloids galantamine and lycorine based on electrochemical simulation in addition to in vivo and in vitro models.
PMID 23176740
Abstract
Alkaloids from the plant family of Amaryllidaceae, such as galantamine (GAL) and lycorine (LYC), are known to exhibit numerous promising biological and pharmacological activities like antibacterial, antiviral or anti-inflammatory effects. Nonetheless, studies on the biotransformation pathway are rare for this substance class, unless approval for use as medication exists. While GAL has become a prescription drug used to alleviate and delay the symptoms of Alzheimer's disease, LYC exhibits potential antitumor properties. However, it has also been linked to toxic effects resulting in nausea and emesis. Whereas there are few publications available describing the metabolic pathway of GAL in animals and humans, the metabolism of LYC is unknown. Therefore, this study is concerned with the investigation of the oxidative metabolism of GAL and LYC, which was achieved by means of three different approaches: electrochemical (EC) simulation coupled on-line to liquid chromatography (LC) with electrospray mass spectrometric (ESI-MS) detection was applied in addition to in vivo experiments in beagle dog analyzing plasma (BP) and in vitro incubations with rat liver microsomes (RLM). This way, it should be investigated if electrochemistry can be used to predict the oxidative metabolism of alkaloids. For GAL, the EC model was capable of predicting most metabolites observed during microsomal and plasma studies, including N-demethylated, dehydrogenated and oxygenated products or a combination of these. LYC was found to be metabolized far less than GAL in the animal-based approaches, but several EC oxidation products were generated. Some principal metabolic routes could successfully be correlated for this alkaloid as well, comprising dehydrogenation, dehydration to ungeremine and oxygenation reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1660
Galanthamine hydrobromide from Lycoris sp., ≥94% (HPLC)
C17H21NO3 · HBr
L5139
Lycorine hydrochloride, ≥98% (TLC), powder
C16H17NO4 · HCl