EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and biological evaluation of glucose-containing scutellarein derivatives as neuroprotective agents based on metabolic mechanism of scutellarin in vivo.
PMID 23177255
Abstract
Based on metabolic mechanism of scutellarin in vivo that scutellarin could be hydrolyzed into scutellarein by β-glucuronide enzyme, some glucose-containing scutellarein derivatives were designed and synthesized through the introduction of glucose moiety at C-7 position of scutellarein via a glucosidic bond. Biological activity evaluation showed that these glucose-containing scutellarein derivatives exhibited potent DPPH radical scavenging activities. Furthermore, the improvement of physicochemical properties such as anticoagulant and neuroprotective activities alongside with the water solubility was achieved by introducing glucose. These findings suggest that the introduction of the glucose moiety to scutellarein wattants further development of this kind of compounds as neuroprotective agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0327
Scutellarein, ≥98% (HPLC)
C15H10O6
73577
Scutellarin, analytical standard
C21H18O12