EMAIL THIS PAGE TO A FRIEND
Otolaryngologic clinics of North America
Anesthesia for office procedures.
PMID 23177401
Abstract
The purpose of this article is to provide otolaryngologists with specific instructions on how to adequately perform topical anesthesia for the most commonly performed laryngeal office-based procedures. In this article, patient selection, lidocaine dosing and safety, and patient monitoring are reviewed.