EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
The solution structure of rabbit IgG accounts for its interactions with the Fc receptor and complement C1q and its conformational stability.
PMID 23178865
Abstract
Solution structures for antibodies are critical to understand function and therapeutic applications. The stability of the solution structure of rabbit IgG in different buffers and temperatures was determined by analytical ultracentrifugation and X-ray and neutron scattering. Rabbit IgG showed a principally monomeric species, which is well resolved from small amounts of a dimeric species. The proportion of dimer increased with increased concentration, decreased temperature and heavy water from 8% to 25% in all buffers except for high salt (250 mM NaCl). The Guinier X-ray radius of gyration R(G) likewise increased with concentration in 137 mM NaCl buffer but was unchanged in 250 mM NaCl buffer. The Guinier neutron R(G) values increased as the temperature decreased. The X-ray and neutron distance distribution curves P(r) revealed two peaks, M1 and M2, whose positions did not change with concentration to indicate unchanged structures under all these conditions. The maximum dimension increased with concentration because of dimer formation. Constrained scattering modeling reproducibly revealed very similar asymmetric solution structures for monomeric rabbit IgG in different buffers, in which the Fab-Fc and Fab-Fab pairs were separated by maximally extended hinge structures. The dimer was best modeled by two pairs of Fab regions forming tip-to-tip contacts. The intact rabbit IgG structures explained the ability of its two ligands, the Fc receptor and complement C1q, to bind to the top of its Fc region that is fully accessible and unhindered by the Fab regions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1740 Complement component C1q from human serum, ≥95% (SDS-PAGE)