EMAIL THIS PAGE TO A FRIEND
International journal of antimicrobial agents
The novel polymyxin derivative NAB739 is remarkably less cytotoxic than polymyxin B and colistin to human kidney proximal tubular cells.
PMID 23182536