EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Wood pulp as an immobilization matrix for the continuous production of isopropanol and butanol.
PMID 23184175
Abstract
The study was focused on developing a continuous method to produce an alcohol mixture suitable to be used as a gasoline supplement. The immobilized column reactor with wood pulp fibers was successfully used for the continuous production of butanol and isopropanol using Clostridium beijerinckii DSM 6423. A sugar mixture (glucose, mannose, galactose, arabinose and xylose) representing lignocellulose hydrolysate was used as a substrate for the production of solvents. The effect of dilution rate on solvent production was studied during continuous operation. The maximum total solvent concentration of 11.99 g/l was obtained at a dilution rate of 0.16 h(-1). The maximum solvent productivity (5.58 g/l h) was obtained at a dilution rate of 1.5 h(-1). The maximum solvent yield of 0.45 g/g from sugar mixture was observed at 0.25 h(-1). The system will be further used for the solvent production using wood hydrolysate as a substrate.