EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Piezoelectric-polarization-enhanced photovoltaic performance in depleted-heterojunction quantum-dot solar cells.
PMID 23184468
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372595
Lead(II) sulfide, 99.9% trace metals basis
PbS