EMAIL THIS PAGE TO A FRIEND
Advanced healthcare materials
Cascaded photoinduced drug delivery to cells from multifunctional core-shell mesoporous silica.
PMID 23184746
Abstract
Different bioactive molecules are released into living cells from lipid-covered mesoporous silica nanoparticles. The release is triggered by light, as the particles feature covalently attached photosensitizers as membrane-opening agents. It is demonstrated that the particles achieve endosomal escape and that they release their cargo into the cytosol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258385
Protoporphyrin IX disodium salt, ≥90% (HPLC)
C34H32N4Na2O4