EMAIL THIS PAGE TO A FRIEND
Advanced healthcare materials
Smart interface materials integrated with microfluidics for on-demand local capture and release of cells.
PMID 23184803
Abstract
Stimuli responsive, smart interface materials are integrated with microfluidic technologies creating new functions for a broad range of biological and clinical applications by controlling the material and cell interactions. Local capture and on-demand local release of cells are demonstrated with spatial and temporal control in a microfluidic system.