EMAIL THIS PAGE TO A FRIEND
Advanced healthcare materials
Mimicking the biological ligand-receptor principle for universal target gene delivery mediated by avidin-biotin interaction.
PMID 23184873
Abstract
A facile and flexible targeting gene-delivery strategy is reported by mimicking the biological ligand-receptor principle based on avidin-biotin interaction. In vitro and in vivo investigations demonstrate that this strategy provides a simple, but universal alternative for potential target gene delivery as well as drug delivery.
Related Materials