PloS one
Pan-Bcl-2 inhibitor AT-101 enhances tumor cell killing by EGFR targeted T cells.
PMID 23185240
Abstract
Pancreatic cancer is a deadly disease and has the worst prognosis among almost all cancers and is in dire need of new and improved therapeutic strategies. Conditioning of tumor cells with chemotherapeutic drug has been shown to enhance the anti-tumor effects of cancer vaccines and adoptive cell therapy. In this study, we investigated the immunomodulatory effects of pan-Bcl-2 inhibitor AT-101 on pancreatic cancer (PC) cell cytotoxicity by activated T cells (ATC). The effects of AT-101 on cytotoxicity, early apoptosis, and Granzyme B (GrzB) and IFN-γ signaling pathways were evaluated during EGFR bispecific antibody armed ATC (aATC)-mediated killing of L3.6pl and MiaPaCa-2 PC cells pre-sensitized with AT-101. We found that pretreatment of tumor cells with AT-101 enhanced susceptibility of L3.6pl and MiaPaCa-2 tumor cells to ATC and aATC-mediated cytotoxicity, which was in part mediated via enhanced release of cytolytic granule GrzB from ATC and aATC. AT-101-sensitized L3.6pl cells showed up-regulation of IFN-γ-mediated induction in the phosphorylation of Ser(727)-Stat1 (pS(727)-Stat1), and IFN-γ induced dephosphorylation of phospho-Tyr(705)-Stat3 (pY(705)-Stat3). Priming (conditioning) of PC cells with AT-101 can significantly enhance the anti-tumor activity of EGFRBi armed ATC through increased IFN-γ induced activation of pS(727)-Stat1 and inhibition of pY(705)-Stat3 phosphorylation, and resulting in increased ratio of pro-apoptotic to anti-apoptotic proteins. Our results verify enhanced cytotoxicity after a novel chemotherapy conditioning strategy against PC that warrants further in vivo and clinical investigations.