PloS one
Lactobacillus proteins are associated with the bactericidal activity against E. coli of female genital tract secretions.
PMID 23185346
Abstract
Female genital tract secretions are bactericidal for Escherichia (E.) coli ex vivo. However, the intersubject variability and molecules that contribute to this activity have not been defined. The bactericidal activity and concentration of immune mediators in cervicovaginal lavage (CVL) collected from 99 healthy women were determined. CVL reduced the number of E. coli colonies by 68% [-26, 100] (median [range]). CVL were active against laboratory and clinical isolates of E. coli, but were inactive against Lactobacillus species. Bactericidal activity correlated with the concentration of protein recovered (p<0.001), but not with cytokines, chemokines or antimicrobial peptides. Four CVL with>90% inhibitory activity (active) and two with<30% activity were subjected to MS/MS proteomic analysis. 215 proteins were identified and six were found exclusively in active samples. Four of these corresponded to Lactobacillus crispatus or jensenii proteins. Moreover, culture supernatants from Lactobacillus jensenii were bactericidal for E. coli. Both host and commensal microbiota proteins contribute to mucosal defense. Identification of these proteins will facilitate the development of strategies to maintain a healthy vaginal microbiome and prevent colonization with pathogenic bacteria such as E. coli that increase the risk for urinary tract infections, preterm labor and perinatal infection.