EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Photoaddition of 5-bromouracil to uracil in oligonucleotides leading to 5,5'-bipyrimidine-type adducts: mechanism of the photoreaction.
PMID 23186224
Abstract
5-Bromouracil ((Br)U) modified di- and hexanucleotides having (Br)U flanked on the 5' or the 3' side by uracil (U) have been synthesized, and their photochemical reactivity was examined under the conditions of irradiation with near UV light. The results indicate that the primary photochemical process in all of these compounds involves the formation of an intermediate cyclobutane phodoadduct composed of (Br)U and U, which undergoes further photochemically and thermally induced transformations to 5,5'-bipyrimidine type adducts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852473
5-Bromouracil, 98%
C4H3BrN2O2