EMAIL THIS PAGE TO A FRIEND
Journal of molecular modeling
Cytotoxic effect and molecular docking of 4-ethoxycarbonylmethyl-1-(piperidin-4-ylcarbonyl)-thiosemicarbazide--a novel topoisomerase II inhibitor.
PMID 23187686
Abstract
The preliminary cytotoxic effect of 4-ethoxycarbonylmethyl-1-(piperidin-4-ylcarbonyl)-thiosemicarbazide hydrochloride (1)-a potent topoisomerase II inhibitor-was measured using a MTT assay. It was found that the compound decreased the number of viable cells in both estrogen receptor-positive MCF-7 and estrogen receptor-negative MDA-MB-231breast cancer cells, with IC(50) values of 146 ± 2 and 132 ± 2 μM, respectively. To clarify the molecular basis of the inhibitory action of 1, molecular docking studies were carried out. The results suggest that 1 targets the ATP binding pocket.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89050
Thiosemicarbazide, puriss. p.a., 98%
CH5N3S
T33405
Thiosemicarbazide, 99%
CH5N3S