EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Oligomerization of site-specific nicking endonuclease BspD6I at high protein concentrations].
PMID 23189557
Abstract
Ability of site-specific nickase BspD6I (Nt.BspD6I) to oligomerize at concentrations > or = 0.5 microM (> or = 0.035 mg/mL) is studied. Three states of Nt.BspD6I are registered via electrophoretic studies both in the presence and in the absence of DNA. Estimation of their molecular mass allows assigning them as a monomer, a dimer and a trimer. Both dimeric and monomeric Nt.BspD6I are shown to hydrolyze its DNA substrate with the identical specificity. Calculation of the electrostatic potential distribution on the Nt.BspD6I globule surface shows that the protein molecule is a dipole. The Nt. BspD6I oligomeric forms are likely to be the result of ionic protein interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7291 Deoxyribonuclease I RNase-free solution from bovine pancreas