EMAIL THIS PAGE TO A FRIEND
Hypertension research : official journal of the Japanese Society of Hypertension
Does the PPAR-γ-activating property of telmisartan provide a benefit in clinical practice?
PMID 23190691