EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Simplifying alternating peptide synthesis by protease-catalyzed dipeptide oligomerization.
PMID 23190865
Abstract
A unique route to perfectly alternating oligopeptides is described. First, AG-OEt was prepared by standard chemical coupling. Then, AG-OEt was converted by papain-catalysis in 30 s to (AG)(x) (80%-yield, x = 9.4 ± 0.3). Extension of this strategy to other alternating peptide compositions as well as the importance of selecting a protease and reaction conditions that avoid competing transamidation and hydrolysis reactions is discussed.