EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Design, synthesis, antinociceptive and anti-inflammatory activities of novel piroxicam analogues.
PMID 23192189
Abstract
In this paper we report the design, synthesis, antinociceptive and anti-inflammatory activities of a series of benzothiazine N-acylhydrazones 14a–h, planned by structural modification of piroxicam (1), a non steroidal anti-inflammatory drug. Among the synthesized analogues, compounds 14f (LASSBio-1637) and 14g (LASSBio-1639) were identified as novel antinociceptive and anti-inflammatory prototypes, active by oral administration, acting by a mechanism of action that seems to be different from that of piroxicam, since they were inactive as an inhibitor of cyclooxygenase (COX-1 and COX-2) at concentrations of 10 mM.