EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Transcriptional regulatory factor X6 (Rfx6) increases gastric inhibitory polypeptide (GIP) expression in enteroendocrine K-cells and is involved in GIP hypersecretion in high fat diet-induced obesity.
PMID 23192339
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ABD28
Anti-Rfx6 Antibody, serum, clone, from rabbit