EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Low symmetry pyrazole-based tripodal tetraamine ligands: metal complexes and ligand decomposition reactions.
PMID 23192397
Abstract
The new low symmetry pyrazole-based tripodal tetraamine ligands 2-(1H-pyrazol-1-yl)-N,N-bis(1H-pyrazol-1-ylmethyl)ethanamine (bmpz) and 2-(1H-pyrazol-1-yl)-N-[2-(1H-pyrazol-1-yl)ethyl]-N-(1H-pyrazol-1-ylmethyl)ethanamine (bepz) have been prepared and characterised, as have metal complexes containing these ligands. X-ray crystal structures of [Co(bmpz)Cl](2)[CoCl(4)]·H(2)O, [Co(bmpz)MeCN](ClO(4))(2)·0.13H(2)O, [Zn(bmpz)MeCN](ClO(4))(2)·0.15H(2)O, [Zn(bepz)OH(2)](ClO(4))(2)·0.5H(2)O and [(Co(bepz)Cl)(2)]Cl(2)·6H(2)O confirm coordination of the intact tripodal ligands to the metal ions through all four N atoms. However, attempts to make Cu(2+) complexes containing bmpz and bepz gave, respectively, [Cu(7)Cl(2)]·0.2H(2)O and [Cu(8)Cl(2)] (7 = 1-(1H-pyrazol-1-yl)-N-(1H-pyrazol-1-ylmethyl)ethanamine, 8 = 2-(1H-pyrazol-1-yl)-N-[2-(1H-pyrazol-1-yl)ethyl]ethanamine), complexes containing the tridentate ligands 7 and 8 which are formed by loss of a pyrazolylmethyl arm from the appropriate tripodal ligand. This decomposition reaction occurs in protic solvents both in the presence and absence of metal ions, and is ascribed to the presence of an aminal functionality in the tripodal ligands. A possible mechanism for the decomposition, based on NMR and ESMS data, is suggested.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82620
Pyrazole, purum, ≥98.0% (GC)
C3H4N2
P56607
Pyrazole, 98%
C3H4N2