EMAIL THIS PAGE TO A FRIEND
Schweizer Monatsschrift fur Zahnmedizin = Revue mensuelle suisse d'odonto-stomatologie = Rivista mensile svizzera di odontologia e stomatologia
Fluorides - mode of action and recommendations for use.
PMID 23192605
Abstract
Various authors have shown that the caries decline in the industrialized countries during recent decades is based on the use of fluorides, of which local fluoride application in the form of fluoridated toothpastes is of primary importance. The caries-protective potential of fluorapatite is quite low; in contrast, dissolved fluorides in the vicinity of enamel are effective both in promoting remineralization and inhibiting demineralization. Considering the fact that the caries decline occurred at the same time that local fluoridation measures became widely used, the conclusion seems justified that regular application of F⁻ can inhibit caries.