EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
ATDC5: an excellent in vitro model cell line for skeletal development.
PMID 23192741
Abstract
The ATDC5 cell line is derived from mouse teratocarcinoma cells and characterized as a chondrogenic cell line which goes through a sequential process analogy to chondrocyte differentiation. Thus, it is regarded as a promising in vitro model to study the factors that influence cell behaviors during chondrogenesis. It also provides insights in exploring signaling pathways related to skeletal development as well as interactions with innovative materials. To date, over 200 studies have utilized ATDC5 to obtain lots of significant findings. In this review, we summarized the literature of ATDC5 related studies and emphasized the application of ATDC5 in chondrogenesis. In addition, the general introduction of ATDC5 including its derivation and characterization is covered in this article.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
ATDC5, 99072806