EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Structure of the O-specific polysaccharide from the lipopolysaccharide of Psychrobacter cryohalolentis K5(T) containing a 2,3,4-triacetamido-2,3,4-trideoxy-L-arabinose moiety.
PMID 23193973
Abstract
A novel constituent of bacterial polysaccharides, 2,3,4-triacetamido-2,3,4-trideoxy-L-arabinose, was found in the O-specific polysaccharide from the lipopolysaccharide of Psychrobacter cryohalolentis K5(T) and identified by 1D and 2D (1)H and (13)C NMR studies of the polysaccharide and a disaccharide obtained by solvolysis of the polysaccharide with triflic acid. The following structure of the branched polysaccharide was established by sugar analysis, triflic acid solvolysis, Smith degradation, and 2D NMR spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9524
DL-Arabinose, ≥98%
C5H10O5