Biochimica et biophysica acta
Induction of a trophoblast-like phenotype by hydralazine in the p19 embryonic carcinoma cell line.
PMID 23195226
Chemicals that affect cellular differentiation through epigenetic mechanisms have potential utility in treating a wide range of diseases. Hydralazine decreases DNA methylation in some cell types but its effect on differentiation has not been well explored. After five days of exposure to hydralazine, P19 embryocarcinoma cells displayed a giant cell morphology and were binucleate, indicative of a trophoblast-like morphology. Other trophoblast-like properties included the intermediary filament Troma-1/cytokeratin 8 and the transcription factor Tead4. A decrease in CpG methylation at three sites in the TEAD4 promoter and the B1 repeated sequence was observed. Knocking down expression of Tead4 with siRNA blocked the increase in Troma-1/cytokeratin 8 and over expression of Tead4 induced the expression of Troma-1/cytokeratin 8. Cells treated for 5days with hydralazine were no longer capable of undergoing retinoic acid-mediated neuronal differentiation. An irreversible loss of the pluripotent transcription factor Oct-4 was observed following hydralazine exposure. In summary, hydralazine induces P19 cells to assume a trophoblast-like phenotype by upregulating Tead4 expression through a mechanism involving DNA demethylation.