EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Treatment of phosphogypsum waste produced from phosphate ore processing.
PMID 23195600
Abstract
Phosphogypsum (PG), primary byproduct from phosphoric acid production, is accumulated in large stockpiles and occupies vast areas of land. Phosphogypsum is a technologically enhanced naturally occurring radioactive material (TE-NORM) that contains radionuclides from (238)U and (232)Th decay series which are of most radio-toxicity. The reduction in concentration of radionuclides content from PG was based on leaching of (226)Ra, (210)Pb, (238)U and (40)K using tri-butyl phosphate (TBP) and tri-octyl phosphine oxide (TOPO) in kerosene. The factors which affect the leaching process such as contact time, concentration of the solvent and temperature were optimized. Based on the experimental results, about 92.1, 88.9, 83.4, 94.6% of (226)Ra, (210)Pb, (238)U and (40)K respectively were successfully removed from the PG. The reduction in the concentration of radionuclides was accompanied by reduction in the concentration of rare earth elements (∑REE) equals to 80.1%. Using the desired organic extractant under optimum conditions for treatment of the PG waste leads to obtain a decontaminated product that can be safely used in many industrial applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
255696
Calcium sulfate, ≥99.99% trace metals basis
CaO4S
237132
Calcium sulfate, −325 mesh, 99%
CaO4S
C3771
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
CaO4S · 2H2O
12056
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
CaO4S · 2H2O
255548
Calcium sulfate dihydrate, ACS reagent, 98%
CaO4S · 2H2O
12090
Calcium sulfate hemihydrate, purum, ≥97.0%
CaO4S · 0.5H2O
238961
Drierite, with indicator, 4 mesh
CaO4S
456071
Drierite, with indicator, 6 mesh
CaO4S
238988
Drierite, with indicator, 8 mesh
CaO4S
238910
Drierite, without indicator, 4 mesh
CaO4S
238937
Drierite, without indicator, 8 mesh
CaO4S
737828
Drierite, with indicator, 10-20 mesh
CaO4S
Y0000279
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
C12H27O4P
00675
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
C12H27O4P
90820
Tributyl phosphate, puriss., ≥99.0% (GC)
C12H27O4P
158615
Tributyl phosphate, 97%
C12H27O4P
240494
Tributyl phosphate, ≥99%
C12H27O4P
34108
Tributyl phosphate, analytical standard
C12H27O4P
PHR1205
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
C12H27O4P
223301
Trioctylphosphine oxide, ReagentPlus®, 99%
C24H51OP
92848
Trioctylphosphine oxide, Selectophore
C24H51OP
00676
Trioctylphosphine oxide, for extraction analysis, ≥98.5%
C24H51OP
346187
Trioctylphosphine oxide, technical grade, 90%
C24H51OP