EMAIL THIS PAGE TO A FRIEND
World journal of surgery
Mean (standard deviation) or mean (standard error of mean): time to ponder.
PMID 23196340
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8396
Piperacillin sodium salt, penicillin analog
C23H26N5NaO7S